2-Azido-1,3-dimethylimidazolinium hexafluorophosphate

Supplier: Matrix Scientific

Synonyms: 2-Azido-1,3-dimethylimidazolinium Hexafluorophosphate, 2-Azido-4,5-dihydro-1,3-dimethyl-1H-imidazolium hexafluorophosphate

070675-1G 070675-5G
101945-866EA 233.85 USD
101945-866 101945-868
2-Azido-1,3-dimethylimidazolinium hexafluorophosphate
2-Azido-1,3-dimethylimidazolinium hexafluorophosphate

Hazard: Irritant

Formula: C₅H₁₀F₆N₅P
MDL Number: MFCD18782891
CAS Number: 1266134-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR