2,9-Dioxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070673-100MG 070673-250MG 070673-500MG
101945-854EA 989.23 USD
101945-854 101945-856 101945-858
2,9-Dioxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester ≥95%
2,9-Dioxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD18782890
CAS Number: 1272412-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR