4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine Hydrochloride, 4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride hydrochloride, (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine hydrochloride

070672-500MG 070672-1G
101945-850EA 509.23 USD
101945-850 101945-852
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥95%
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂S
MW: 185.6700 g/mol
MDL Number: MFCD11114559
CAS Number: 116529-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR