tert-Butyl-N-(1,1-dioxothian-4-yl)carbamate

Supplier: Matrix Scientific

070671-250MG 070671-500MG 070671-1G
101945-842EA 640 USD
101945-842 101945-844 101945-846
tert-Butyl-N-(1,1-dioxothian-4-yl)carbamate
tert-Butyl-N-(1,1-dioxothian-4-yl)carbamate

Hazard: Irritant

Formula: C₁₀H₁₉NO₄S
MDL Number: MFCD18839237
CAS Number: 595597-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR