3-Amino-2,2,4,4-tetramethylthietane hydrochloride ≥95%

Supplier: Matrix Scientific

070667-250MG 070667-500MG 070667-1G
101945-824EA 524.62 USD
101945-824 101945-826 101945-828
3-Amino-2,2,4,4-tetramethylthietane hydrochloride ≥95%
3-Amino-2,2,4,4-tetramethylthietane hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNS
MDL Number: MFCD18839236
CAS Number: 80875-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR