2,2,4,4-Tetramethylthietan-3-one ≥96%

Supplier: Matrix Scientific

070666-250MG 070666-500MG 070666-1G
101945-818EA 524.62 USD
101945-818 101945-820 101945-822
2,2,4,4-Tetramethylthietan-3-one ≥96%
2,2,4,4-Tetramethylthietan-3-one

Hazard: Irritant

Formula: C₇H₁₂OS
MDL Number: MFCD09865540
CAS Number: 58721-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR