Spiro[3.3]heptane-2,6-dicarboxylicacid monomethyl ester ≥96%

Supplier: Matrix Scientific

Synonyms: 6-(Methoxycarbonyl)spiro[3.3]heptane-2-carboxylic acid

070659-500MG 070659-1G
101945-786EA 1396.92 USD
101945-786 101945-788
Spiro[3.3]heptane-2,6-dicarboxylicacid monomethyl ester ≥96%
Spiro[3.3]heptane-2,6-dicarboxylicacid monomethyl ester

Hazard: Irritant

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD13195611
CAS Number: 10481-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR