6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-(Boc-amino)spiro[3.3]heptane-2-carboxylate, Methyl 6-((tert-butoxycarbonyl)amino)spiro[3.3]heptane-2-carboxylate

070658-500MG 070658-1G
101945-782EA 1801.54 USD
101945-782 101945-784
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester ≥95%
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₄H₂₃NO₄
MW: 269.34 g/mol
MDL Number: MFCD18425636
CAS Number: 170508-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR