6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6-((tert-Butoxycarbonyl)amino)spiro[3.3]heptane-2-carboxylic acid, 6-{[(tert-butoxy)carbonyl]amino}spiro[3.3]heptane-2-carboxylic acid
070657-250MG
070657-500MG
070657-1G
101945-776EA
1396.92
USD
101945-776
101945-778
101945-780
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid ≥95%
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₂₁NO₄ MW: 255.32 g/mol |
MDL Number:
MFCD11099888 CAS Number: 1087798-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...