(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl N-{6-hydroxyspiro[3.3]heptan-2-yl}carbamate, 2-(Boc-amino)-6-hydroxyspiro[3.3]heptane, tert-Butyl (6-hydroxyspiro[3.3]heptan-2-yl)carbamate

070656-250MG 070656-500MG 070656-1G
101945-770EA 2384.62 USD
101945-770 101945-772 101945-774
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥95%
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD09702342
CAS Number: 1000933-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR