2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate ≥95%

Supplier: Matrix Scientific

070653-100MG 070653-250MG 070653-500MG
101945-754EA 1221.54 USD
101945-754 101945-756 101945-758
2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate ≥95%
2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate

Hazard: Irritant

Formula: C₇H₁₁NO₂·CF₃CO₂H
MW: 255.191 g/mol
MDL Number: MFCD19982773
CAS Number: 1523572-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR