6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6,6-difluoro-2-azaspiro[3.3]heptane-2-carboxylate

070651-100MG 070651-250MG 070651-500MG
101945-742EA 1221.54 USD
101945-742 101945-744 101945-746
6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%
6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₇F₂NO₂
MW: 233.26 g/mol
MDL Number: MFCD18432776
CAS Number: 1264635-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR