7-Fluoro-1H-indazole ≥95%

Supplier: Matrix Scientific

070648-500MG 070648-1G
101945-728EA 524.62 USD
101945-728 101945-730
7-Fluoro-1H-indazole ≥95%
7-Fluoro-1H-indazole

Hazard: Irritant

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
MDL Number: MFCD09026976
CAS Number: 341-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR