4-Chloropyrrolo[1,2-f][1,2,4]triazine ≥96%

Supplier: Matrix Scientific

070645-500MG 070645-1G
101945-716EA 1338.46 USD
101945-716 101945-718
4-Chloropyrrolo[1,2-f][1,2,4]triazine ≥96%
4-Chloropyrrolo[1,2-f][1,2,4]triazine

Hazard: Irritant

Formula: C₆H₄ClN₃
MDL Number: MFCD11519387
CAS Number: 888720-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR