5-Methyl-3-(methylsulfonyl)-1,2,4-triazine

Supplier: Matrix Scientific

070643-500MG 070643-1G 070643-5G
101945-710EA 930.77 USD
101945-710 101945-712 101945-714
5-Methyl-3-(methylsulfonyl)-1,2,4-triazine
5-Methyl-3-(methylsulfonyl)-1,2,4-triazine

Hazard: Irritant

Formula: C₅H₇N₃O₂S
MW: 173.19 g/mol
MDL Number: MFCD18325192
CAS Number: 945915-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR