Benzylhydrazine monohydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Benzylhydrazine hydrochloride

070641-5G 070641-25G
101945-700EA 138.46 USD
101945-700 101945-702
Benzylhydrazine monohydrochloride ≥95%
Benzylhydrazine monohydrochloride

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 158.63 g/mol
MDL Number: MFCD01722685
CAS Number: 1073-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR