2-Bromo-6-chlorothieno[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

070640-1G 070640-5G
101945-694EA 930.77 USD
101945-694 101945-696
2-Bromo-6-chlorothieno[2,3-b]pyridine ≥95%
2-Bromo-6-chlorothieno[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₃BrClNS
MDL Number: MFCD08706311
CAS Number: 68236-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR