O-2-Propynylhydroxylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-(Prop-2-yn-1-yl)hydroxylamine hydrochloride

070638-250MG 070638-1G
101945-686EA 1221.54 USD
101945-686 101945-688
O-2-Propynylhydroxylamine hydrochloride ≥95%
O-2-Propynylhydroxylamine hydrochloride

Hazard: Irritant

Formula: C₃H₆ClNO
MW: 107.54 g/mol
MDL Number: MFCD12912207
CAS Number: 21663-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR