5-Iodo-3-methylisothiazole 95

Supplier: Matrix Scientific

070637-250MG 070637-1G
101945-682EA 1687.69 USD
101945-682 101945-684
5-Iodo-3-methylisothiazole 95
5-Iodo-3-methylisothiazole

Hazard: Irritant

Formula: C₄H₄INS
MW: 225.05 g/mol
MDL Number: MFCD17012839
CAS Number: 20067-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR