4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-7-yl)-1,3,2-dioxaborolane
Supplier: Matrix Scientific
070636-250MG
070636-1G
101945-678EA
655.05
USD
101945-678
101945-680
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-7-yl)-1,3,2-dioxaborolane
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-7-yl)-1,3,2-dioxaborolane
Hazard: Irritant/Refrigerate
Formula:
C₁₄H₂₃BO₄ |
MDL Number:
MFCD11110349 CAS Number: 1049730-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...