4-Bromo-3-methylisothiazole 95

Supplier: Matrix Scientific

070632-250MG 070632-1G
101945-662EA 1192.31 USD
101945-662 101945-664
4-Bromo-3-methylisothiazole 95
4-Bromo-3-methylisothiazole

Hazard: Irritant

Formula: C₄H₄BrNS
MW: 178.06 g/mol
MDL Number: MFCD11110311
CAS Number: 930-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR