7-Bromo-3,4-dihydro-2H-benzo[b]oxepin-5-one ≥95%
Supplier: Matrix Scientific
070629-1G
070629-5G
101945-650EA
815.38
USD
101945-650
101945-652
7-Bromo-3,4-dihydro-2H-benzo[b]oxepin-5-one ≥95%
7-Bromo-3,4-dihydro-2H-benzo[b]oxepin-5-one
Hazard: Irritant
Formula:
C₁₀H₉BrO₂ |
MDL Number:
MFCD08446940 CAS Number: 55580-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...