1'-(tert-Butoxycarbonyl)-2,3-dihydrospiro-[indene-1,4'-piperidine]-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
070625-250MG
070625-500MG
101945-628EA
815.38
USD
101945-628
101945-630
1'-(tert-Butoxycarbonyl)-2,3-dihydrospiro-[indene-1,4'-piperidine]-3-carboxylic acid ≥97%
1'-(tert-Butoxycarbonyl)-2,3-dihydrospiro-[indene-1,4'-piperidine]-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₉H₂₅NO₄ |
MDL Number:
MFCD08461259 CAS Number: 185526-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...