tert-Butyl-2-oxo-1,7-diazaspiro[4.4]nonane-7-carboxylate ≥97%

Supplier: Matrix Scientific

070619-250MG 070619-500MG
101945-598EA 653.85 USD
101945-598 101945-600
tert-Butyl-2-oxo-1,7-diazaspiro[4.4]nonane-7-carboxylate ≥97%
tert-Butyl-2-oxo-1,7-diazaspiro[4.4]nonane-7-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD12198506
CAS Number: 1160246-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR