tert-Butyl-6-methoxy-3-oxo-2,3-dihydrospiro[indene-1,4'-piperidine]-1'-carboxylate ≥97%

Supplier: Matrix Scientific

070617-250MG 070617-500MG
101945-590EA 1687.69 USD
101945-590 101945-592
tert-Butyl-6-methoxy-3-oxo-2,3-dihydrospiro[indene-1,4'-piperidine]-1'-carboxylate ≥97%
tert-Butyl-6-methoxy-3-oxo-2,3-dihydrospiro[indene-1,4'-piperidine]-1'-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₅NO₄
MW: 331.42 g/mol
MDL Number: MFCD08460794
CAS Number: 910442-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR