7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 7-[(tert-butoxy)carbonyl]-7-azaspiro[3.5]nonane-2-carboxylic acid

070615-250MG 070615-500MG 070615-1G
101945-578EA 1687.69 USD
101945-578 101945-580 101945-582
7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid ≥96%
7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₃NO₄
MW: 269.34 g/mol
MDL Number: MFCD12198556
CAS Number: 873924-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR