4-(Hydroxyimino)methylphenylboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-((Hydroxyimino)methyl)phenyl)boronic acid
070611-1G
070611-5G
101945-562EA
175.38
USD
101945-562
101945-564
4-(Hydroxyimino)methylphenylboronic acid ≥95%
4-(Hydroxyimino)methylphenylboronic acid
Hazard: Irritant
Formula:
C₇H₈BNO₃ MW: 164.96 g/mol |
MDL Number:
MFCD04039325 CAS Number: 179942-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...