Thietan-3-ylcarbamic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl thietan-3-ylcarbamate

070608-250MG 070608-1G
101945-558EA 640 USD
101945-558 101945-560
Thietan-3-ylcarbamic acid tert-butyl ester ≥95%
Thietan-3-ylcarbamic acid tert-butyl ester

Hazard: Irritant

Formula: C₈H₁₅NO₂S
MW: 189.28 g/mol
MDL Number: MFCD18325185
CAS Number: 943437-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR