4-Isoquinolinol ≥95%

Supplier: Matrix Scientific

Synonyms: Isoquinolin-4-ol, 4-Hydroxyisoquinoline

070607-1G 070607-5G
101945-554EA 287.69 USD
101945-554 101945-556
4-Isoquinolinol ≥95%
4-Isoquinolinol

Hazard: Irritant

Formula: C₉H₇NO
MW: 145.1580 g/mol
MDL Number: MFCD00234514
CAS Number: 3336-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR