Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

070603-250MG 070603-500MG
101945-536EA 2850.77 USD
101945-536 101945-538
Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid ≥95%
Trans-2-Benzyloxycarbonylaminocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MW: 249.266 g/mol
MDL Number: MFCD03844619
CAS Number: 1212272-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR