cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

070602-250MG 070602-500MG
101945-532EA 2850.77 USD
101945-532 101945-534
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid ≥95%
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD03844615
CAS Number: 685508-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR