1-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070599-250MG 070599-500MG
101945-516EA 2675.38 USD
101945-516 101945-518
1-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%
1-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD17016205
CAS Number: 1100748-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR