2-Phenyl-2,6-diazaspiro[3.3]heptan-1-one ≥95%

Supplier: Matrix Scientific

070598-250MG 070598-500MG
101945-512EA 2675.38 USD
101945-512 101945-514
2-Phenyl-2,6-diazaspiro[3.3]heptan-1-one ≥95%
2-Phenyl-2,6-diazaspiro[3.3]heptan-1-one

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD13180714
CAS Number: 960079-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR