2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate(2:1)

070596-250MG 070596-500MG 070596-1G
101945-500EA 930.77 USD
101945-500 101945-502 101945-504
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate ≥97%
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate

Hazard: Irritant

Formula: C₂₂H₃₈N₄O₈
MW: 486.57 g/mol
MDL Number: MFCD12404932
CAS Number: 1041026-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR