3-Amino-2,2-dimethylcyclobutanecarboxylic acid ≥97%

Supplier: Matrix Scientific

070595-250MG 070595-500MG 070595-1G
101945-494EA 1396.92 USD
101945-494 101945-496 101945-498
3-Amino-2,2-dimethylcyclobutanecarboxylic acid ≥97%
3-Amino-2,2-dimethylcyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD19441794
CAS Number: 783260-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR