tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-oxo-2-azaspiro[3.3]heptane-2-carboxylate

070594-250MG 070594-1G
101945-488EA 703.08 USD
101945-488 101945-492
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate ≥95%
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MW: 211.2576 g/mol
MDL Number: MFCD15071430
CAS Number: 1181816-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR