2-Boc-6-amino-2-azaspiro[3.3]heptane ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-azaspiro[3.3]heptane, N2-BOC protected, tert-Butyl 6-amino-2-azaspiro[3.3]heptane-2-carboxylate

070593-250MG 070593-500MG 070593-1G
101945-482EA 1512.31 USD
101945-482 101945-484 101945-486
2-Boc-6-amino-2-azaspiro[3.3]heptane ≥95%
2-Boc-6-amino-2-azaspiro[3.3]heptane

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.2900 g/mol
MDL Number: MFCD15071432
CAS Number: 1211586-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR