tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-azaspiro[3.3]heptan-6-ylcarbamate, tert-butyl N-{2-azaspiro[3.3]heptan-6-yl}carbamate

070592-250MG 070592-500MG 070592-1G
101945-476EA 1512.31 USD
101945-476 101945-478 101945-480
tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate ≥95%
tert-Butyl-2-azaspiro[3.3]hept-6-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD11858159
CAS Number: 1118786-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR