N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl N-{6-aminospiro[3.3]heptan-2-yl}carbamate, tert-Butyl (6-aminospiro[3.3]heptan-2-yl)carbamate, 2,6-Diaminospiro[3.3]heptane, 2-BOC protected

070591-250MG 070591-500MG 070591-1G
101945-470EA 1512.31 USD
101945-470 101945-472 101945-474
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester ≥95%
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD09054707
CAS Number: 1239589-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR