1,4-Diazabicyclo[4.3.1]decan-2-one

Supplier: Matrix Scientific

070589-250MG 070589-500MG 070589-1G
101945-460EA 1512.31 USD
101945-460 101945-462 101945-464
1,4-Diazabicyclo[4.3.1]decan-2-one
1,4-Diazabicyclo[4.3.1]decan-2-one

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD18325182
CAS Number: 1000577-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR