Thietan-3-one ≥95%

Supplier: Matrix Scientific

070585-1G 070585-5G
101945-448EA 153.85 USD
101945-448 101945-450
Thietan-3-one ≥95%
Thietan-3-one

Hazard: Irritant

Formula: C₃H₄OS
Melting Pt: 69…72 °C
MDL Number: MFCD11848696
CAS Number: 22131-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR