4-(Tributylstannyl)quinoline ≥94%

Supplier: Matrix Scientific

070582-1G 070582-5G
101945-434EA 756.92 USD
101945-434 101945-436
4-(Tributylstannyl)quinoline ≥94%
4-(Tributylstannyl)quinoline

Hazard: Irritant

Formula: C₂₁H₃₃NSn
MDL Number: MFCD18325178
CAS Number: 1272412-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR