2-(7-Octyn-1-yl)-1H-isoindole-1,3-dione ≥96%
Supplier: Matrix Scientific
070581-1G
070581-5G
101945-430EA
1106.15
USD
101945-430
101945-432
2-(7-Octyn-1-yl)-1H-isoindole-1,3-dione ≥96%
2-(7-Octyn-1-yl)-1H-isoindole-1,3-dione
Hazard: Irritant
Formula:
C₁₆H₁₇NO₂ |
MDL Number:
MFCD18325177 CAS Number: 17170-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...