1,9-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070578-250MG 070578-500MG
101945-416EA 1687.69 USD
101945-416 101945-418
1,9-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester ≥95%
1,9-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD12406520
CAS Number: 1158750-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR