1,8-Diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1,8-diazaspiro[5.5]undecane-8-carboxylate

070576-250MG 070576-500MG
101945-406EA 1687.69 USD
101945-406 101945-408
1,8-Diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester ≥95%
1,8-Diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD11223538
CAS Number: 1086394-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR