2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2,8-diazaspiro[5.5]undecane-2-carboxylate

070575-250MG 070575-1G
101945-402EA 655.05 USD
101945-402 101945-404
2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥95%
2,8-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD09701315
CAS Number: 954240-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR