2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070574-100MG 070574-250MG 070574-500MG
101945-396EA 815.38 USD
101945-396 101945-398 101945-400
2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester ≥95%
2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MDL Number: MFCD11223510
CAS Number: 189333-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR