9-Oxa-2-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester ≥95%
Supplier: Matrix Scientific
070572-250MG
070572-500MG
101945-386EA
1396.92
USD
101945-386
101945-388
9-Oxa-2-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester ≥95%
9-Oxa-2-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₂₅NO₃ |
MDL Number:
MFCD18325173 CAS Number: 1259489-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...