2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate, 9-Boc-2,9-diazaspiro[5.5]undecane

070569-250MG 070569-1G
101945-372EA 75.38 USD
101945-372 101945-374
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥95%
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester

Hazard: Irritant/Refrigerate

Formula: C₁₄H₂₆N₂O₂
Boiling Pt: 354 °C
Density: 1.05
Flash Pt: 168 °C
MDL Number: MFCD10700117
CAS Number: 1023595-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR