3-Oxa-9-azaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070568-250MG 070568-1G
101945-368EA 930.77 USD
101945-368 101945-370
3-Oxa-9-azaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥95%
3-Oxa-9-azaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₅NO₃
MDL Number: MFCD18325172
CAS Number: 1259489-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR